Learn More
Medchemexpress LLC Rhodamine 123 | 62669-70-9 | MFCD00012664 | 99.6% | 380.82 | C21H17ClN2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD081610MG
Encompass_Preferred
Rhodamine 123 is a green-fluorescent xanthene dye used as a probe of mitochondrial membrane potential and as a general fluorescent tracer in cell biology and bioanalytical assays. It is supplied as a stable solid for preparation of stock solutions for microscopy, flow cytometry, and fluorescence spectroscopy.
  • High purity for reproducible fluorescence.
  • Cell-permeable cationic dye that accumulates in mitochondria.
  • Suitable for microscopy, flow cytometry, and plate-reader assays.
  • Stable solid form for convenient storage and stock solution preparation.
  • Compatible with common fluorescent imaging and spectroscopic workflows.
Catalog No. 50-002-97647