Learn More
Medchemexpress LLC HY-D0309 1g Medchemexpress, Rhodamine 6G CAS:989-38-8 Purity:>98%
SDP
Catalog No. 501933169
Encompass_Preferred
Medchemexpress, HY-D0309 1g Rhodamine 6G CAS:989-38-8 Rhodamine 6G is a rhodamine analog useful in Pgp efflux assays. It can be used in characterizing the kinetics of MRP1- mediated efflux, and as a laser dye and potential mitochondrial probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3169 Supplier Medchemexpress LLC Supplier No. HYD03091G
Medchemexpress, HY-D0309 1g Rhodamine 6G CAS:989-38-8 Rhodamine 6G is a rhodamine analog useful in Pgp efflux assays. It can be used in characterizing the kinetics of MRP1- mediated efflux, and as a laser dye and potential mitochondrial probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.