Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhodamine 6G

Catalog No. 252433250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-243-3250MG 250 mg
25-243-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-243-3250MG Supplier Sigma Aldrich Supplier No. 252433250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rhodamine 6G (R6G) is an organic laser dye and can be used to study the probes as it has a high quantum yield for fluorescence. It can be used as a fluorescence tracker which helps in defining the spectroscopic characteristics for achieving a high conversion efficiency and precision of measurements. The absorption of R6G in different solvents can range between 400-700 nm.R6G can be used as a cationic dye to determine the photocatalytic ability of TiO2 reinforced graphene oxide composites that can be potentially used in self-cleaning applications. It can be used to calculate the quantum yield of glutathione-capped copper nanoclusters (CuNCs) for fluorescent thermo-responsive nanomaterials and gold nanoparticles (AuNPs) for fluorescent/X-ray computed tomography.

Specifications

CAS 989-38-8
Molecular Formula C28H30N2O3 -+ HCl
Linear Formula C28H31N2O3Cl
Quantity 250 mg
MDL Number MFCD00012665
Synonym Basic Red 1
Molecular Weight (g/mol) 479.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.