Learn More
TARGETMOL CHEMICALS INC RHODAMINE 6G 5000MG
SDP
Catalog No. 502577329
Encompass_Preferred
Also available in 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rhodamine 6G (Basic Red 1) is a rhodamine analog useful in Pgp efflux assays. It can be widely used in characterizing the kinetics of MRP1- mediated efflux as a laser dye and potential mitochondrial probe. purity: 99%
Catalog No. 50-257-7329 Supplier TARGETMOL CHEMICALS INC Supplier No. T167445G
Also available in 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rhodamine 6G (Basic Red 1) is a rhodamine analog useful in Pgp efflux assays. It can be widely used in characterizing the kinetics of MRP1- mediated efflux as a laser dye and potential mitochondrial probe. purity: 99%