Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine 6G, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111016010 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6010 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6010 Supplier MilliporeSigma Supelco Supplier No. 5622625MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 989-38-8
Physical Form Neat
Linear Formula C28H31N2O3 Cl
MDL Number MFCD00012665
Synonym Basic Red 1
Type Analytical Standard
Quantity 25 mg
Formula Weight 479.01
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.