Learn More
Medchemexpress LLC Rhodamine 6G | 989-38-8 | 98.0% | 250 MG
SDP
Catalog No. 5000414439
Encompass_Preferred
Rhodamine dyes are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials. They attach to mitochondria, producing bright fluorescence. At certain concentrations, rhodamine dyes exhibit low toxicity to cells, making them commonly used for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe
  • Specifically recognizes mitochondrial membrane potentials
  • Attaches to mitochondria and produces bright fluorescence
  • Low toxicity to cells at certain concentrations
  • Commonly used to detect mitochondria in animal cells, plant cells, and microorganisms
  • Prolongs survival and improves clinical parameters in in vivo studies
  • Inhibits tumor growth and metastases in melanoma-transplanted mice
  • Enters the circulatory system and labels leukocytes
Catalog No. 50-004-14439 Supplier Medchemexpress LLC Supplier No. 250MGHYD0309
Rhodamine dyes are membrane-permeable cationic fluorescent probes that specifically recognize mitochondrial membrane potentials. They attach to mitochondria, producing bright fluorescence. At certain concentrations, rhodamine dyes exhibit low toxicity to cells, making them commonly used for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Membrane-permeable cationic fluorescent probe
  • Specifically recognizes mitochondrial membrane potentials
  • Attaches to mitochondria and produces bright fluorescence
  • Low toxicity to cells at certain concentrations
  • Commonly used to detect mitochondria in animal cells, plant cells, and microorganisms
  • Prolongs survival and improves clinical parameters in in vivo studies
  • Inhibits tumor growth and metastases in melanoma-transplanted mice
  • Enters the circulatory system and labels leukocytes