Learn More
AdipoGen Rhodamine 6G bis AEA amide bis
SDP
Catalog No. 502560127
Encompass_Preferred
Chemical. CAS 1173097-37-4. Formula C30H37N5O2 . C4H2F6O4. MW 727.69. Synthetic. Fluorescent labelling.
Catalog No. 50-256-0127 Supplier AdipoGen Supplier No. CDXR0025M250
Chemical. CAS 1173097-37-4. Formula C30H37N5O2 . C4H2F6O4. MW 727.69. Synthetic. Fluorescent labelling.