Learn More
AdipoGen Rhodamine 6G hexanediamine ami
SDP
Catalog No. 502560136
Encompass_Preferred
Chemical. CAS 1140505-40-3. Formula C32H40N4O2 . C4H2F6O4. MW 740.73. Synthetic. N-6-Aminohexylrhodamine 6G-amide bistrifluoroacetate is a rhodamine 6G-amide derivative useful as a bioreagent in fluorescence assays.
Catalog No. 50-256-0136 Supplier AdipoGen Supplier No. CDXR0021M010
Chemical. CAS 1140505-40-3. Formula C32H40N4O2 . C4H2F6O4. MW 740.73. Synthetic. N-6-Aminohexylrhodamine 6G-amide bistrifluoroacetate is a rhodamine 6G-amide derivative useful as a bioreagent in fluorescence assays.