Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhodamine 6G hydrazide | 932013-08-6 | 98.1% | 428.53 g/mol | C26H28N4O2 | 10 MG
SDP

Catalog No. 5000298850
Encompass_Preferred

Rhodamine 6G hydrazide is a xanthene-based fluorescent dye supplied as a research reagent for use in selective colorimetric and fluorescent sensing, imaging, and labeling applications.

  • Fluorescent xanthene dye used for colorimetric and fluorescent sensing.
  • High purity (98.05%) as stated on the product datasheet.
  • Molecular weight 428.53 g/mol; formula C26H28N4O2.
  • Supplied as a solid powder in multiple milligram pack sizes suitable for laboratory use.
  • CAS number 932013-08-6 for unambiguous identification.
  • Intended for research use only; not for clinical or diagnostic use.

Catalog No. 50-002-98850 Supplier Medchemexpress LLC Supplier No. HYD159910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhodamine 6G hydrazide is a xanthene-based fluorescent dye supplied as a research reagent for use in selective colorimetric and fluorescent sensing, imaging, and labeling applications.

  • Fluorescent xanthene dye used for colorimetric and fluorescent sensing.
  • High purity (98.05%) as stated on the product datasheet.
  • Molecular weight 428.53 g/mol; formula C26H28N4O2.
  • Supplied as a solid powder in multiple milligram pack sizes suitable for laboratory use.
  • CAS number 932013-08-6 for unambiguous identification.
  • Intended for research use only; not for clinical or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.