Learn More
AdipoGen Rhodamine 6G p-DAX amide bis
SDP
Catalog No. 502560130
Encompass_Preferred
Chemical. CAS 591742-78-8. Formula C34H36N4O2 . C4H2F6O4. MW 760.72. Synthetic. Fluorescent labelling.
Catalog No. 50-256-0130 Supplier AdipoGen Supplier No. CDXR0024M050
Chemical. CAS 591742-78-8. Formula C34H36N4O2 . C4H2F6O4. MW 760.72. Synthetic. Fluorescent labelling.