Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rhodamine B

Catalog No. p-7021294
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1357218 50 g
AAA1357230 250 g
AAA135720B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1357218 Supplier Thermo Scientific Chemicals Supplier No. A1357218
Only null left
Add to Cart

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

Solubility
Soluble in water, benzene and ethanol. Slightly soluble in acetone, hydrochloric acid, sodium hydroxide, methyl cellosolve and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point ∼210°C (decomposition)
Beilstein 4119648
Merck Index 14,8183
Solubility Information Soluble in water,benzene and ethanol. Slightly soluble in acetone,hydrochloric acid,sodium hydroxide,methyl cellosolve and alcohol.
Formula Weight 479.02
Density 1.31
Quantity 50 g
Chemical Name or Material Rhodamine B
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 201-383-9
RTECSNumber BP3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.