Learn More
Selleck Chemical LLC Rhodamine B
SDP
Catalog No. 5000003045
Encompass_Preferred
Rhodamine B is used as a tracer dye in water to determine the rate and direction of flow and transport It is a staining fluorescent dye used in fluorescence microscopy flow cytometry fluorescence correlation spectroscopy and ELISA in biotechnology fields
Catalog No. 50-000-03045 Supplier Selleck Chemical LLC Supplier No. S564125MG
Rhodamine B is used as a tracer dye in water to determine the rate and direction of flow and transport It is a staining fluorescent dye used in fluorescence microscopy flow cytometry fluorescence correlation spectroscopy and ELISA in biotechnology fields