Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine B, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015615 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5615 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5615 Supplier MilliporeSigma Supelco Supplier No. 7975425MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhodamine B is a cationic dye.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 81-88-9
Physical Form Neat
Linear Formula C28H31 ClN2O3
MDL Number MFCD00011931
Melting Point 210-211
Synonym Basic Violet 10; Brilliant Pink B; Rhodamine O; Tetraethylrhodamine
Type Analytical Standard
Quantity 25 mg
Formula Weight 479.01
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.