Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine B, Fisher Chemical

Catalog No. R21100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21-100
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

For tungsten and antimony determinations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym [9-(o-Carboxyphenyl)-6-(diethylamino)-3H-xanthen-3-ylidene] Diethyl-ammonium Chloride, Tetraethylrhodamine
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name 9-(2-carboxyphenyl)-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point 210°C
Identification Pass Test
Formula Weight 479
Packaging Glass Bottle
Color Index No. 45170
Color Green
Quantity 100 g
Physical Form Solid
Chemical Name or Material Rhodamine B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.