Learn More
Medchemexpress LLC Rhodamine B hydrazide | 74317-53-6 | MFCD07808762 | 99.9% | 456.58 | C28H32N4O2 | 10MM 1ML
SDP
Catalog No. 5000236002
Encompass_Preferred
Rhodamine B hydrazide is a red fluorescent hydrazide dye supplied as a solid or as a 10 mM solution in DMSO. It is used for labeling carbonyl-containing biomolecules and as a fluorescent probe for detection of metal ions and reactive species, with conventional excitation/emission around 510/578 nm.
  • Red fluorescence with excitation/emission approximately 510/578 nm.
  • Hydrazide functional group enables conjugation to aldehydes and ketones.
  • Available as solid or ready-to-use 10 mM solution in DMSO for convenience.
  • High purity (99.9%) suitable for analytical and labeling applications.
  • Store protected from light; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-36002 Supplier Medchemexpress LLC Supplier No. HY12364510MM 1ML
Add to Cart
Add to Cart
Rhodamine B hydrazide is a red fluorescent hydrazide dye supplied as a solid or as a 10 mM solution in DMSO. It is used for labeling carbonyl-containing biomolecules and as a fluorescent probe for detection of metal ions and reactive species, with conventional excitation/emission around 510/578 nm.
  • Red fluorescence with excitation/emission approximately 510/578 nm.
  • Hydrazide functional group enables conjugation to aldehydes and ketones.
  • Available as solid or ready-to-use 10 mM solution in DMSO for convenience.
  • High purity (99.9%) suitable for analytical and labeling applications.
  • Store protected from light; in solvent, store at -80°C for long-term stability.