Learn More
Medchemexpress LLC Rhodamine B hydrazide | 74317-53-6 | MFCD07808762 | 99.9% | 456.58 g/mol | C28H32N4O2 | 100MG
SDP
Catalog No. 5000236001
Encompass_Preferred
Rhodamine B hydrazide is a rhodamine-derived fluorescent probe for labeling and detection in fluorescence assays. It is supplied as an off-white to yellow solid, is soluble in DMSO, and exhibits conventional excitation/emission near 510/578 nm.
  • Fluorescent probe with excitation/emission 510/578 nm.
  • High purity (99.9%).
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Solid, off-white to yellow appearance.
  • Available in multiple package sizes, including 100 MG.
Catalog No. 50-002-36001 Supplier Medchemexpress LLC Supplier No. HY123645100MG
Add to Cart
Add to Cart
Rhodamine B hydrazide is a rhodamine-derived fluorescent probe for labeling and detection in fluorescence assays. It is supplied as an off-white to yellow solid, is soluble in DMSO, and exhibits conventional excitation/emission near 510/578 nm.
  • Fluorescent probe with excitation/emission 510/578 nm.
  • High purity (99.9%).
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Solid, off-white to yellow appearance.
  • Available in multiple package sizes, including 100 MG.