Learn More
Apexbio Technology LLC Rhodamine B isothiocyanate 36877-69-7 50mg
SDP
Catalog No. 5000064062
Encompass_Preferred
Rhodamine B isothiocyanate (RBITC) is a fluorescent dye bearing an isothiocyanate functional group enabling its covalent conjugation specifically to amino groups in proteins and peptides Upon excitation by green light wavelengths this compound emits characteristic orange-red fluorescence allowing for visualization and detection of labeled biomolecules in cellular and tissue samples Due to its stable covalent binding to target proteins RBITC is employed extensively in fluorescence microscopy immunohistochemical analysis and in vitro labeling studies in disciplines such as cell biology neuroscience and immunology Its chemical structure facilitates selective attachment aiding the detailed investigation of cellular localization protein dynamics and molecular interactions in biological systems
Catalog No. 50-000-64062 Supplier Apexbio Technology LLC Supplier No. B879150
Rhodamine B isothiocyanate (RBITC) is a fluorescent dye bearing an isothiocyanate functional group enabling its covalent conjugation specifically to amino groups in proteins and peptides Upon excitation by green light wavelengths this compound emits characteristic orange-red fluorescence allowing for visualization and detection of labeled biomolecules in cellular and tissue samples Due to its stable covalent binding to target proteins RBITC is employed extensively in fluorescence microscopy immunohistochemical analysis and in vitro labeling studies in disciplines such as cell biology neuroscience and immunology Its chemical structure facilitates selective attachment aiding the detailed investigation of cellular localization protein dynamics and molecular interactions in biological systems