Learn More
Rhodamine B Isothiocyanate-Dextran, Sigma-Aldrich
Catalog No. 01258905 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
01-258-905 100 mg
1 options
Catalog No. 01-258-905 Supplier MilliporeSigma Supelco Supplier No. R8881100MG
Add to Cart
Add to Cart
Rhodamine B isothiocyanate-Dextran is a fluorescent probe for endocytosis and perfusion studies.
  • Suitable for investigating cellular endocytotic activity.
  • Can be used to track marked cells and study perfusion in animals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mg
Product Type Fluorescence Reagent
Label Type Other Labels or Dyes
Color Dark Purple