Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine B Isothiocyanate, Sigma-Aldrich

Catalog No. 01258918 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-918 500 mg
01-258-935 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-918 Supplier MilliporeSigma Supelco Supplier No. 283924500MG
Add to Cart
Add to Cart

Rhodamine B isothiocyanate (RBITC) is a fluorescent dye. It acts as an anterograde and retrograde marker of retinal neurons in the adult rat. (current GD)It is also known as RBITC, RhIc, RITC, and TRITC.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36877-69-7

Specifications

Quantity 500 mg
Product Type Fluorescence Reagent
Label Type Other Labels or Dyes
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.