Learn More
Medchemexpress LLC Rhodamine B NHS ester | 144700-55-0 | 96.9% | 576.08 | C32H34ClN3O5 | 10 MG
SDP
Catalog No. 5000297680
Encompass_Preferred
Rhodamine B NHS ester is an N-hydroxysuccinimide (NHS) activated fluorescent dye for covalent labeling of primary amines in proteins, peptides, and other biomolecules. It forms stable amide bonds to introduce a rhodamine fluorophore for fluorescence microscopy, flow cytometry, and biochemical assays.
  • NHS-activated for efficient and selective amine coupling.
  • Bright rhodamine fluorescence suitable for microscopy and flow cytometry.
  • Molecular weight 576.08 g/mol and formula C32H34ClN3O5.
  • High purity (~96.9%) for reliable labeling performance.
  • Available in small research pack sizes for convenient use.
Catalog No. 50-002-97680 Supplier Medchemexpress LLC Supplier No. HYD236710MG
Rhodamine B NHS ester is an N-hydroxysuccinimide (NHS) activated fluorescent dye for covalent labeling of primary amines in proteins, peptides, and other biomolecules. It forms stable amide bonds to introduce a rhodamine fluorophore for fluorescence microscopy, flow cytometry, and biochemical assays.
  • NHS-activated for efficient and selective amine coupling.
  • Bright rhodamine fluorescence suitable for microscopy and flow cytometry.
  • Molecular weight 576.08 g/mol and formula C32H34ClN3O5.
  • High purity (~96.9%) for reliable labeling performance.
  • Available in small research pack sizes for convenient use.