Learn More
Abcam Rhodamine B octadecyl ester perchlorate, Lipophilic energy transfer acceptor, 20MG
Catalog No. 502705263
Encompass_Preferred
MW 795.49 g/mol, Purity >90%. Lipophilic energy transfer acceptor from lipophilic fluoresceins in fluorescence energy transfer assays (FRET) for cell fusion.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5263 Supplier Abcam Supplier No. AB14536420MG
MW 795.49 g/mol, Purity >90%. Lipophilic energy transfer acceptor from lipophilic fluoresceins in fluorescence energy transfer assays (FRET) for cell fusion.
The product is subject to the following: Abcam Restricted Use Statement