Learn More
Medchemexpress LLC Rhodamine B PEG2-NH2 | 95.3% | 566.13 | 25 MG
SDP
Catalog No. 5000348648
Encompass_Preferred
Rhodamine B PEG2-NH2 is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. For research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
Catalog No. 50-003-48648 Supplier Medchemexpress LLC Supplier No. 25MGHY138396
Rhodamine B PEG2-NH2 is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. For research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation