Learn More
Medchemexpress LLC Rhodamine B PEG2-NH2 | 95.3% | 566.13 | 50 MG
SDP
Catalog No. 5000350993
Encompass_Preferred
Rhodamine B PEG2-NH2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50993 Supplier Medchemexpress LLC Supplier No. HY13839650MG
Rhodamine B PEG2-NH2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins