Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine B, Sigma-Aldrich

Catalog No. 01259035 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-035 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-035 Supplier MilliporeSigma Supelco Supplier No. R6626500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhodamine B has been used:

  • to represent fluorescence of tetra methyl rhodamine (TMR)
  • in the preparation of adhesive resin for confocal laser scanning microscopy
  • as a fluorescent dye in intrahemolymph dye dosing and retinal image acquisition
  • to depict brain-specific dye capture
  • for imaging targets
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Weight (g/mol) 479.01 g/mol

Specifications

Color Green
Quantity 500 g
Percent Purity ≥95%
Chemical Name or Material Rhodamine B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.