Learn More
Rhodamine B Solution, 0.2% in isopropanol, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 81-88-9 |
| Linear Formula | C28H31 ClN2O3 |
| Formula Weight | 479.01 |
| Grade | TLC |
| Concentration or Composition (by Analyte or Components) | 0.2% in isopropanol |
| Density | 0.79 g/mL (at 20°C) |
| Quantity | 100 mL |
| Flash Point | 12°C - Closed Cup |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.