Learn More
Abcam Rhodamine Conjugation Kit (Fast) - Lightning-Link, 3 x 100UG
Catalog No. 502590111
Encompass_Preferred
Rhodamine conjugation / labeling in < 20 mins with 30 secs hands-on time using Rhodamine Conjugation Kit (Fast) - Lightning-Link ab188286. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0111 Supplier Abcam Supplier No. AB1882863X100UG
Rhodamine conjugation / labeling in < 20 mins with 30 secs hands-on time using Rhodamine Conjugation Kit (Fast) - Lightning-Link ab188286. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement