Learn More
Cayman Chemical Rhodamine dithenoyl hydrazide
SDP
Catalog No. 502989463
Encompass_Preferred
A fluorescent and colorimetric probe for the detection of ferric iron; fluoresces upon iron binding; ex/em = 534/550-700 nm; emits fluorescence at 588 nm in the presence of ferric iron but not ferrous iron or in a panel of 11 additional cations and 10 anions; exhibits a visual shift from colorless to pink upon addition of ferric iron to aqueous solutions or live cells, which can be detected colorimetrically
Catalog No. 50-298-9463 Supplier Cayman Chemical Supplier No. 310955 MG
A fluorescent and colorimetric probe for the detection of ferric iron; fluoresces upon iron binding; ex/em = 534/550-700 nm; emits fluorescence at 588 nm in the presence of ferric iron but not ferrous iron or in a panel of 11 additional cations and 10 anions; exhibits a visual shift from colorless to pink upon addition of ferric iron to aqueous solutions or live cells, which can be detected colorimetrically