Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhodamine Green™-X, Succinimidyl Ester, Hydrochloride, mixed isomers

Catalog No. R6113
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6113 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6113 Supplier Invitrogen™ Supplier No. R6113
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rhodamine Green™-X, Succinimidyl Ester, Hydrochloride, mixed isomers

The amine-reactive Rhodamine Green™-X, succinimidyl ester can be used to create green-fluorescent bioconjugates with excitation/emission maxima ∼502/527 nm. This dye is the nonsulfonated analog of the Alexa Fluor™ 488 dye.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Rhodamine Green™
Label Type Classic Dyes
Product Line Rhodamine Green
Emission 527
Excitation 502
Show More Show Less
What is the excitation and emission wavelength for rhodamine?

Rhodamine is a generic term for a wide variety of cationic dyes whose fluorescence emission can range from green, orange to red. The table below lists the excitation and emission maxima (nm), as well as molar extinction coefficients (“EC”; cm-1 M-1), for various rhodamine dyes (data derived with dye dissolved in methanol).

Dye Excitation Emission EC
Rhodamine B 568 583 88,000
Rhodamine 123 507 529 101,000
Rhodamine 110 499 521 92,000
Rhodamine 6G 528 551 105,000
XRITC 572 596 92,000

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.