Learn More
Medchemexpress LLC Rhodamine-N3 chloride | 2363751-90-8 | 99.9% | 847.44 | 5 MG
SDP
Catalog No. 5000429043
Encompass_Preferred
Rhodamine-N3 chloride is an azide-rhodamine fluorescent dye utilized for labeling biomolecules that contain alkyne groups. It functions as a click chemistry reagent, possessing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups, exhibiting an excitation/emission (Ex/Em) profile of 544/576 nm.
  • Azide-rhodamine fluorescent dye
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Excitation/emission (Ex/Em) = 544/576 nm
  • Enables labeling of biomolecules with alkyne groups
  • Versatile for various click chemistry applications
Catalog No. 50-004-29043 Supplier Medchemexpress LLC Supplier No. HYD12695MG
Rhodamine-N3 chloride is an azide-rhodamine fluorescent dye utilized for labeling biomolecules that contain alkyne groups. It functions as a click chemistry reagent, possessing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups, exhibiting an excitation/emission (Ex/Em) profile of 544/576 nm.
  • Azide-rhodamine fluorescent dye
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition reactions
  • Excitation/emission (Ex/Em) = 544/576 nm
  • Enables labeling of biomolecules with alkyne groups
  • Versatile for various click chemistry applications