Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhodamine-N3 chloride | 2363751-90-8 | 99.9% | 847.44 g·mol⁻¹ | C44H59ClN8O7 | 10 MG
SDP

Catalog No. 5000297303
Encompass_Preferred

Rhodamine-N3 chloride is an azide-functionalized rhodamine fluorescent dye used to label alkyne-containing biomolecules via click chemistry. It delivers strong fluorescence (excitation 544 nm, emission 576 nm) and is supplied as a solid chloride salt for research applications.

  • Azide-functional rhodamine dye for click chemistry labeling.
  • Excitation 544 nm, emission 576 nm for reliable fluorescence detection.
  • High purity (≈99.85%) and molecular weight 847.44 g·mol⁻¹.
  • Solid, brown to black appearance; compatible with common organic solvents.
  • Store at 4 °C, sealed, protected from moisture and light.
  • Supplied as a 10 mg vial for small-scale labeling experiments.

Catalog No. 50-002-97303 Supplier Medchemexpress LLC Supplier No. HYD126910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhodamine-N3 chloride is an azide-functionalized rhodamine fluorescent dye used to label alkyne-containing biomolecules via click chemistry. It delivers strong fluorescence (excitation 544 nm, emission 576 nm) and is supplied as a solid chloride salt for research applications.

  • Azide-functional rhodamine dye for click chemistry labeling.
  • Excitation 544 nm, emission 576 nm for reliable fluorescence detection.
  • High purity (≈99.85%) and molecular weight 847.44 g·mol⁻¹.
  • Solid, brown to black appearance; compatible with common organic solvents.
  • Store at 4 °C, sealed, protected from moisture and light.
  • Supplied as a 10 mg vial for small-scale labeling experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.