Learn More
Biotium Rhodamine Phalloidin, 300U
Catalog No. 501964057
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. Red fluorescent rhodamine phalloidin has excitation/emission at 540/565 nm.
Catalog No. 50-196-4057 Supplier Biotium Supplier No. 00027
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. Red fluorescent rhodamine phalloidin has excitation/emission at 540/565 nm.