Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodanile Blue, Thermo Scientific™

Catalog No. AAA1362203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1362203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1362203 Supplier Thermo Scientific Chemicals Supplier No. A1362203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14969-56-3
Molecular Formula C48H48ClN5O3
MDL Number MFCD00011937

Specifications

Formula Weight 778.4
Quantity 1 g
Chemical Name or Material Rhodanile Blue
Hazard Category H319-H351
Hazard Statement GHS H Statement
H351-H319
Suspected of causing cancer.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 239-040-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.