Learn More
Medchemexpress LLC Rhodanine | 141-84-4 | 98.0% | 133.20 | 250 G
SDP
Catalog No. 5000453309
Encompass_Preferred
Rhodanine is a selective Echovirus 12 proliferation inhibitor and intermediate. It is used to determine tannase activity and can be utilized in the synthesis of anticancer and antibacterial agents.
  • Selective Echovirus 12 proliferation inhibitor.
  • Used to determine tannase activity.
  • Can be utilized in the synthesis of anticancer and antibacterial agents.
  • Significantly inhibits echovirus 12 in primary monkey kidney cell cultures.
Catalog No. 50-004-53309 Supplier Medchemexpress LLC Supplier No. 250GHYW010365
Rhodanine is a selective Echovirus 12 proliferation inhibitor and intermediate. It is used to determine tannase activity and can be utilized in the synthesis of anticancer and antibacterial agents.
  • Selective Echovirus 12 proliferation inhibitor.
  • Used to determine tannase activity.
  • Can be utilized in the synthesis of anticancer and antibacterial agents.
  • Significantly inhibits echovirus 12 in primary monkey kidney cell cultures.