Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodanine 99.0+%, TCI America™
SDP

Catalog No. R0017500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0017500G 500 g
R001725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0017500G Supplier TCI America Supplier No. R0017500G

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
MDL Number MFCD00005488
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
Synonym rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazoline
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

Melting Point 168°C
Color Yellow
Quantity 500 g
Formula Weight 133.18
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Rhodanine
EINECSNumber (5)-3410
RTECSNumber VI7700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.