Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodinyl Isovalerate, MP Biomedicals

Catalog No. ICN225569 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225569 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225569 Supplier MP Biomedicals, Inc Supplier No. 0522556980

Chemical Identifiers

CAS 7778-96-3
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
InChI Key OZAWINZSOFVOBJ-AWEZNQCLSA-N
Synonym unii-ry3fd3joga, ry3fd3joga, rhodinyl isovalerianate, 3s-3,7-dimethyloct-7-en-1-yl 3-methylbutanoate, s-3,7-dimethyloct-7-enyl isovalerate, rhodinyl isopentanoate, 3,7-dimethyl-7-octen-1-yl 3-methylbutanoate, rhodinyl 3-methylbutanoate, butanoic acid, 3-methyl-, 3s-3,7-dimethyl-7-octenyl ester, fema no. 2987
PubChem CID 25113547
IUPAC Name [(3S)-3,7-dimethyloct-7-enyl] 3-methylbutanoate
SMILES CC(C)CC(=O)OCCC(C)CCCC(=C)C

Specifications

Quantity 100 g
Chemical Name or Material Rhodinyl Isovalerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.