Learn More
Cayman Chemical Rosarin, Purity: ≥98%, CAS: 84954-93-8, 10 mg
SDP
Catalog No. 502912517
Encompass_Preferred
A glycoside with anti-inflammatory and immune regulatory activities; prevents LPS-induced increases in iNOS, TNF-α, IL-1β, and IL-6 levels in BV-2 microglia at 10 and 50 µg/ml; inhibits concanavalin A- or anti-CD3 and anti-CD28 antibody-induced growth of Jurkat T cells (IC50s = 60 and 68 µM, respectively); increases TRAIL levels in isolated mouse splenic T cells
Catalog No. 50-291-2517 Supplier Cayman Chemical Supplier No. 3521410 MG
A glycoside with anti-inflammatory and immune regulatory activities; prevents LPS-induced increases in iNOS, TNF-α, IL-1β, and IL-6 levels in BV-2 microglia at 10 and 50 µg/ml; inhibits concanavalin A- or anti-CD3 and anti-CD28 antibody-induced growth of Jurkat T cells (IC50s = 60 and 68 µM, respectively); increases TRAIL levels in isolated mouse splenic T cells