Learn More
Cayman Chemical RhodIola rosea RosavIn 50mg
SDP
Catalog No. 502942379
Encompass_Preferred
A diglycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 5.86 UG/ml); inhibits the proliferation of NCI H69, NCI H526, and NCI H446 SCLC cells at 5-100 µM; inhibits concanavalin A- or anti-CD3 and anti-CD28 antibody-induced growth of Jurkat T cells (IC50s = 80 and 74 µM, respectively) and reduces TRAIL levels in isolated mouse splenic T cells at 0.5, 5, and 50 µM; reduces ferroptosis-related mitochondrial changes and increased levels of 4-HNE and MDA in type II alveolar epithelial cells, as well as reduces lung tissue iron levels, in a rat model of intratracheal PM2.5-induced lung injury at 100 µM
Catalog No. 50-294-2379 Supplier Cayman Chemical Supplier No. 3983950 MG
A diglycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 5.86 UG/ml); inhibits the proliferation of NCI H69, NCI H526, and NCI H446 SCLC cells at 5-100 µM; inhibits concanavalin A- or anti-CD3 and anti-CD28 antibody-induced growth of Jurkat T cells (IC50s = 80 and 74 µM, respectively) and reduces TRAIL levels in isolated mouse splenic T cells at 0.5, 5, and 50 µM; reduces ferroptosis-related mitochondrial changes and increased levels of 4-HNE and MDA in type II alveolar epithelial cells, as well as reduces lung tissue iron levels, in a rat model of intratracheal PM2.5-induced lung injury at 100 µM