Learn More
Cayman Chemical Rosin, Purity: ≥98%, CAS: 85026-55-7, 25 mg
SDP
Catalog No. 502946635
Encompass_Preferred
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice
Catalog No. 50-294-6635 Supplier Cayman Chemical Supplier No. 3438325 MG
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice