Learn More
Medchemexpress LLC HY-N0241 5mg Medchemexpress, Rhodionin CAS:85571-15-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN02415MG
Encompass_Preferred
Medchemexpress, HY-N0241 5mg Rhodionin CAS:85571-15-9 Rhodionin, isolated from the root of Rhodiola crenulata, is a specific non-competitive cytochrome P450 2D6 inhibitor with an IC50 of 0.761 μM and a Ki of 0.769 μM. Rhodionin exhibits potent, dose-dependent inhibitory effects on acetylcholinesterase (AChE) with IC50 ranged from 57.50 to 2.43 μg/mL. Rhodionin exhibits potent DPPH free radical scavenging activities, with an IC50 of 19.49 μM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0239