Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium, 0.5% on 3mm (0.12in) alumina pellets, Thermo Scientific™

Catalog No. AA4250718
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA4250718 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4250718 Supplier Thermo Scientific Chemicals Supplier No. 4250718

Hydrogenation of aromatics

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-16-6
Quantity 50g
Molecular Formula Rh
MDL Number MFCD00011201
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
Solubility Information Insoluble in water.
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES [Rh]
IUPAC Name rhodium
Molecular Weight (g/mol) 102.906
PubChem CID 23948
ChEBI CHEBI:33359
Odor Odorless
Chemical Name or Material Rhodium, 0.5% on alumina pellets, 3mm (0.12 in.) alumina pellets
Show More Show Less
EINECSNumber 231-125-0
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.