Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium on activated carbon, 5% Rh, reduced, dry, Thermo Scientific™

Catalog No. AC396490010
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC396490010 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC396490010 Supplier Thermo Scientific Chemicals Supplier No. 396490010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-16-6, 7440-44-0
Molecular Formula Rh
MDL Number MFCD00011201
Quantity 1g
Merck Index 15, 8310
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES [Rh]
IUPAC Name rhodium
Molecular Weight (g/mol) 102.906
PubChem CID 23948
ChEBI CHEBI:33359
Formula Weight 102.91
Packaging Glass bottle
Form Powder
Chemical Name or Material Rhodium on activated carbon
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable solid.
Precautionary Statement GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.