Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rhodium(II) Acetate Dimer

Catalog No. 2090581G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
2 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-905-81G 1 g
20-905-8250MG 250 mg
20-905-82G 2 g
20-905-825G 25 g
20-905-850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Warning
Catalog No. 20-905-81G Supplier Sigma Aldrich Supplier No. 2090581G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in an efficient synthesis of +â-hydroxy-+a-arylacrylates involving the decomposition of diazoetster intermediates with concomitant 1,2-arylmigration.

Specifications

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Linear Formula Rh2(OOCCH3)4
Quantity 1 g
MDL Number MFCD00003538
Synonym Rh2(OAc)4; Dirhodium tetraacetate; Tetrakis(acetato)dirhodium(II)
Molecular Weight (g/mol) 441.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.