Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(II) acetate, dimer, 98+%

Catalog No. AAL15152MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15152MC 100 mg
AAL15152ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15152MC Supplier Thermo Scientific Chemicals Supplier No. L15152MC
Add to Cart

CAS: 15956-28-2 | C8H12O8Rh2 | 441.99 g/mol

Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ?-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving ɑ-diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

Solubility
Soluble in aqueous solution like water. Slightly soluble in ethanol

Notes
Stable under normal conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
PubChem CID 122130469
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 205°C
Quantity 100 mg
Linear Formula [Rh(CO2CH3)2]2
Solubility Information Soluble in aqueous solution like water. Slightly soluble in ethanol
IUPAC Name bis(λ²-rhodium(2+)) tetraacetate
Formula Weight 441.99
Percent Purity ≥98%
Odor Odorless
Chemical Name or Material Rhodium(II) acetate, dimer
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 240-084-8
RTECSNumber VI9361000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.