Learn More
Strem, An Ascensus Company CAS# 31126-95-1. 500mg. Rhodium(II) trifluoroacetate dimer, min. 95%. MFCD00209611
SDP
Catalog No. 5090112251
Encompass
CAS# 31126-95-1. 500mg. Rhodium(II) trifluoroacetate dimer, min. 95%. MFCD00209611. Molecular Weight: 657.88. Molecular Formula: Rh2(OOCCF3)4. Color/form: green pwdr. Strem# 45-1960. http://www.strem.com/catalog/v/45-1960/
Catalog No. 50-901-12251 Supplier Strem, An Ascensus Company Supplier No. 451960500MG
CAS# 31126-95-1. 500mg. Rhodium(II) trifluoroacetate dimer, min. 95%. MFCD00209611. Molecular Weight: 657.88. Molecular Formula: Rh2(OOCCF3)4. Color/form: green pwdr. Strem# 45-1960. http://www.strem.com/catalog/v/45-1960/