Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD15071082 | 1355.14 | C80H60O8Rh2 | 1 G
SDP
Catalog No. 5000301557
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dirhodium(II) carboxylate complex used as a catalyst in organic synthesis and biochemical research. The complex contains two rhodium centers bridged by four triphenylacetate ligands and is commonly applied in carbene-transfer, C-H insertion, and related transformations. Handle under inert gas and protect from light.
Catalog No. 50-003-01557 Supplier Medchemexpress LLC Supplier No. HYW0338311G
Rhodium(II) triphenylacetate dimer is a dirhodium(II) carboxylate complex used as a catalyst in organic synthesis and biochemical research. The complex contains two rhodium centers bridged by four triphenylacetate ligands and is commonly applied in carbene-transfer, C-H insertion, and related transformations. Handle under inert gas and protect from light.