Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD15071082 | 1355.14 | C80H60O8Rh2 | 25 MG
SDP
Catalog No. 5000303833
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dimeric rhodium complex used as a catalyst in organic synthesis, notably for C-H activation and carbene-transfer reactions. It is supplied as a light-sensitive solid and should be protected from light and stored under inert atmosphere at low temperature for optimal stability.
  • Catalyzes C-H activation and carbene-transfer reactions with high selectivity.
  • Improves reaction rates and selectivity in complex molecule construction.
  • Provided as a light-sensitive solid; protect from light during handling and storage.
  • Maintain under inert atmosphere and low temperature for long-term stability.
  • Suitable for research use and synthetic methodology development.
Catalog No. 50-003-03833 Supplier Medchemexpress LLC Supplier No. 25MGHYW033831
Rhodium(II) triphenylacetate dimer is a dimeric rhodium complex used as a catalyst in organic synthesis, notably for C-H activation and carbene-transfer reactions. It is supplied as a light-sensitive solid and should be protected from light and stored under inert atmosphere at low temperature for optimal stability.
  • Catalyzes C-H activation and carbene-transfer reactions with high selectivity.
  • Improves reaction rates and selectivity in complex molecule construction.
  • Provided as a light-sensitive solid; protect from light during handling and storage.
  • Maintain under inert atmosphere and low temperature for long-term stability.
  • Suitable for research use and synthetic methodology development.