Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD15071082 | 1355.14 | C80H60O8Rh2 | 250 MG
SDP
Catalog No. 5000303545
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dirhodium(II) carboxylate complex used as a homogeneous catalyst in organic synthesis. It promotes C-H activation and related transformations, helping to construct complex and biologically active molecules by improving reaction rates and selectivity.
  • Catalyzes C-H activation and related functionalization reactions.
  • Improves reaction rate and selectivity in complex molecule synthesis.
  • Dirhodium(II) carboxylate structure suited for homogeneous catalysis.
  • Available in multiple package sizes for bench-scale experiments.
Catalog No. 50-003-03545 Supplier Medchemexpress LLC Supplier No. 250MGHYW033831
Rhodium(II) triphenylacetate dimer is a dirhodium(II) carboxylate complex used as a homogeneous catalyst in organic synthesis. It promotes C-H activation and related transformations, helping to construct complex and biologically active molecules by improving reaction rates and selectivity.
  • Catalyzes C-H activation and related functionalization reactions.
  • Improves reaction rate and selectivity in complex molecule synthesis.
  • Dirhodium(II) carboxylate structure suited for homogeneous catalysis.
  • Available in multiple package sizes for bench-scale experiments.