Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD15071082 | 1355.14 g/mol | C80H60O8Rh2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW033831100MG
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dirhodium catalyst used in organic synthesis to promote C-H activation, cyclopropanation, and related transformations. Supplied as a solid for laboratory research, it is employed in small-scale catalytic screening and preparative reactions where selective C-H insertion and high reaction rates are required.
  • High catalytic activity for C-H activation and insertion reactions.
  • Useful for cyclopropanation and construction of complex molecules.
  • Supplied as a solid, colorless to light green, suitable for laboratory use.
  • Store protected from light and under inert gas to maintain stability.
  • Accompanied by technical documentation and safety data for handling.
Catalog No. 50-003-01558