Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD15071082 | >95.0% | 1355.14 | C80H60O8Rh2 | 10 MG
SDP
Catalog No. 5000299592
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dirhodium(II) carboxylate complex used as a catalyst and reagent in organic synthesis and biochemical research (CAS 142214-04-8). It is supplied as a solid, typically colorless to light green, and is used in carbene transfer, C-H insertion, and related catalytic transformations. Purity documentation is available from the supplier.
  • Dirhodium metal complex suited for catalytic applications in organic synthesis.
  • Facilitates carbene transfer and C-H insertion reactions.
  • Supplied as a solid with colorless to light green appearance for laboratory use.
  • Offered in multiple small-scale quantities to support research workflows.
  • Documentation and purity information available upon request for quality control.
Catalog No. 50-002-99592 Supplier Medchemexpress LLC Supplier No. HYW03383110MG
Rhodium(II) triphenylacetate dimer is a dirhodium(II) carboxylate complex used as a catalyst and reagent in organic synthesis and biochemical research (CAS 142214-04-8). It is supplied as a solid, typically colorless to light green, and is used in carbene transfer, C-H insertion, and related catalytic transformations. Purity documentation is available from the supplier.
  • Dirhodium metal complex suited for catalytic applications in organic synthesis.
  • Facilitates carbene transfer and C-H insertion reactions.
  • Supplied as a solid with colorless to light green appearance for laboratory use.
  • Offered in multiple small-scale quantities to support research workflows.
  • Documentation and purity information available upon request for quality control.