Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD16875675 | 1355.14 | C80H60O8Rh2 | 50 MG
SDP
Catalog No. 5000301774
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dirhodium carboxylate complex used as a homogeneous catalyst in organic synthesis. It is commonly employed for C-H activation, carbene transfer, and cyclopropanation reactions, and is supplied for research-scale applications.
  • Efficient catalyst for C-H activation and insertion reactions.
  • High selectivity and reaction rate in typical catalytic transformations.
  • Useful for cyclopropanation and carbene-transfer methodologies.
  • Suitable for small-scale research and method development.
  • Provided as a solid reagent compatible with common organic solvents.
Catalog No. 50-003-01774 Supplier Medchemexpress LLC Supplier No. HYW03383150MG
Rhodium(II) triphenylacetate dimer is a dirhodium carboxylate complex used as a homogeneous catalyst in organic synthesis. It is commonly employed for C-H activation, carbene transfer, and cyclopropanation reactions, and is supplied for research-scale applications.
  • Efficient catalyst for C-H activation and insertion reactions.
  • High selectivity and reaction rate in typical catalytic transformations.
  • Useful for cyclopropanation and carbene-transfer methodologies.
  • Suitable for small-scale research and method development.
  • Provided as a solid reagent compatible with common organic solvents.